Methyl-4-(1,3-oxazol-5-yl)benzenecarboxylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 4-(1,3-oxazol-5-yl)benzoate, Methyl 4-(5-Oxazolyl)benzoate
A453194-250MG
A453194-25G
A453194-5G
77004-336EA
18.04
USD
77004-336
77004-338
77004-340
Methyl-4-(1,3-oxazol-5-yl)benzenecarboxylate 98%
Methyl-4-(1,3-oxazol-5-yl)benzenecarboxylate
Precaución: for Research Use Only
Formula:
C₁₁H₉NO₃ MW: 203.2 g/mol |
MDL Number:
MFCD03012557 CAS Number: 179057-14-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...