4-Chloro-2,6-difluorobenzoic acid 98%

Supplier: Ambeed
Warning

A453172-10G A453172-1G A453172-5G
77034-120EA 139 USD
77034-120 77034-122 77034-124
4-Chloro-2,6-difluorobenzoic acid 98%
4-Chloro-2,6-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃ClF₂O₂
MDL Number: MFCD06203570
CAS Number: 196194-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR