4-(4-Bromophenyl)dibenzothiophene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(4-Bromophenyl)dibenzo[b,d]thiophene
A453093-1G
A453093-5G
77016-778EA
19.1
USD
77016-778
77016-780
4-(4-Bromophenyl)dibenzothiophene 98%
4-(4-Bromophenyl)dibenzothiophene
Precaución: for Research Use Only
Formula:
C₁₈H₁₁BrS MW: 339.25 g/mol |
MDL Number:
MFCD19441307 CAS Number: 530402-77-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...