(3-Bromo-2-ethoxy-5-fluorophenyl)boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-2-ethoxy-5-fluorophenylboronic acid

A452855-1G A452855-250MG A452855-5G
77016-772EA 71.09 USD
77016-772 77016-774 77016-776
(3-Bromo-2-ethoxy-5-fluorophenyl)boronic acid 95%
(3-Bromo-2-ethoxy-5-fluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BBrFO₃
MW: 262.86 g/mol
MDL Number: MFCD05865158
CAS Number: 352534-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR