tert-Butyl (2-((2-aminoethyl)(methyl)amino)ethyl)carbamate 95%

Supplier: Ambeed
Warning

A452741-100MG A452741-1G A452741-250MG
76990-760EA 93.36 USD
76990-760 76990-762 76990-764
tert-Butyl (2-((2-aminoethyl)(methyl)amino)ethyl)carbamate 95%
tert-Butyl (2-((2-aminoethyl)(methyl)amino)ethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₂₃N₃O₂
MW: 217.30 g/mol
MDL Number: MFCD18831537
CAS Number: 263162-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR