(2R,5R)-2,5-Dimethylmorpholine hydrochloride 95%

Supplier: Ambeed
Warning

A452380-250MG A452380-1G
77016-770EA 108.22 USD
77016-770 77684-237
(2R,5R)-2,5-Dimethylmorpholine hydrochloride 95%
(2R,5R)-2,5-Dimethylmorpholine hydrochloride

Precaución: For research use only.

Formula: C₆H₁₄ClNO
MW: 151.63 g/mol
MDL Number: MFCD22199177
CAS Number: 1639886-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR