6-Iodo-3-methyl-3H-imidazo[4,5-b]pyridine 95%

Supplier: Ambeed
Warning

A452357-100MG A452357-250MG
77004-332EA 167.63 USD
77004-332 77004-334
6-Iodo-3-methyl-3H-imidazo[4,5-b]pyridine 95%
6-Iodo-3-methyl-3H-imidazo[4,5-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₆IN₃
MDL Number: MFCD11857685
CAS Number: 1138444-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR