3-Bromo-5-(dimethoxymethyl)pyridine 95+%
Supplier: Ambeed
|
Warning
|
A452191-100MG
A452191-1G
A452191-250MG
A452191-5G
77004-324EA
21.22
USD
77004-324
77004-326
77004-328
77004-330
3-Bromo-5-(dimethoxymethyl)pyridine 95+%
3-Bromo-5-(dimethoxymethyl)pyridine
Precaución: for Research Use Only
Formula:
C₈H₁₀BrNO₂ |
MDL Number:
MFCD10699139 CAS Number: 163163-79-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...