2-(3-fluoro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Fluoro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Fluoro-4-methoxybenzeneboronic acid pinacol ester

A452188-100MG A452188-1G A452188-250MG A452188-5G
77016-762EA 14.86 USD
77016-762 77016-764 77016-766 77016-768
2-(3-fluoro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%
2-(3-fluoro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₃
MW: 252.092 g/mol
MDL Number: MFCD05863912
CAS Number: 754226-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR