7-Methylpyrazolo[1,5-a]pyridine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A451938-100MG A451938-250MG
77016-758EA 57.29 USD
77016-758 77016-760
7-Methylpyrazolo[1,5-a]pyridine-3-carboxylic acid 95%
7-Methylpyrazolo[1,5-a]pyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.17 g/mol
MDL Number: MFCD16620169
CAS Number: 16205-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR