1-(5-Bromopyrimidin-2-yl)ethanone 98%
Supplier: Ambeed
|
Warning
|
A451830-10G
A451830-1G
A451830-250MG
A451830-5G
A451830-25G
76990-750EA
305
USD
76990-750
76990-752
76990-754
76990-756
77886-192
1-(5-Bromopyrimidin-2-yl)ethanone 98%
1-(5-Bromopyrimidin-2-yl)ethanone
Precaución: for Research Use Only
Formula:
C₆H₅BrN₂O |
MDL Number:
MFCD18909512 CAS Number: 1189169-37-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...