5-Aminovaleric acid hydrochloride 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 5-Aminovaleric Acid Hydrochloride, 5-Aminopentanoic acid, 5-Aminopentanoic acid Hydrochloride, 5-AVACl, Homopiperidinic acid Hydrochloride, 5-Aminopentanoic acid hydrochloride

A451459-25G A451459-1G A451459-5G
76990-742EA 101.85 USD
76990-742 77684-234 77684-235
5-Aminovaleric acid hydrochloride 98%, Ambeed.Inc
5-Aminovaleric acid hydrochloride

Precaución: For research use only.

Formula: H₂N(CH₂)₄CO₂H·HCl
MW: 153.6073 g/mol
MDL Number: MFCD00012919
CAS Number: 627-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR