Methyl 1-Boc-3-methylpiperidine-3-carboxylate 97%

Supplier: Ambeed
Warning

A450772-100MG A450772-1G A450772-250MG A450772-5G
77034-100EA 8 USD
77034-100 77034-102 77034-104 77034-106
Methyl 1-Boc-3-methylpiperidine-3-carboxylate 97%
Methyl 1-Boc-3-methylpiperidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₃NO₄
MW: 257.33 g/mol
MDL Number: MFCD11977502
CAS Number: 888952-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR