[2-[2-(Amino-κN)ethyl]phenyl-κC][bis(1,1-dimethylethyl)[2',4',6'-tris(1-methylethyl)[1,1'-biphenyl]-2-yl]phosphine]chloropalladium 97%
Supplier: Ambeed
|
Warning
|
A449257-1G
A449257-250MG
A449257-5G
A449257-100MG
76990-726EA
91.24
USD
76990-726
76990-728
77613-388
77684-230
[2-[2-(Amino-κN)ethyl]phenyl-κC][bis(1,1-dimethylethyl)[2',4',6'-tris(1-methylethyl)[1,1'-biphenyl]-2-yl]phosphine]chloropalladium 97%
[2-[2-(Amino-κN)ethyl]phenyl-κC][bis(1,1-dimethylethyl)[2',4',6'-tris(1-methylethyl)[1,1'-biphenyl]-2-yl]phosphine]chloropalladium
Precaución: For research use only.
Formula:
C₃₇H₅₅ClNPPd MW: 686.69 g/mol |
MDL Number:
MFCD12911909 CAS Number: 1142811-12-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...