6-Chloro-5-cyanopicolinic acid 95%

Supplier: Ambeed
Warning

A449074-1G A449074-250MG
76990-720EA 77 USD
76990-720 77890-907
6-Chloro-5-cyanopicolinic acid 95%
6-Chloro-5-cyanopicolinic acid

Precaución: for Research Use Only

Formula: C₇H₃ClN₂O₂
MDL Number: MFCD18258363
CAS Number: 53234-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR