9-(4-Iodophenyl)carbazole 98%

Supplier: Ambeed
Warning

Synonyms: 9-(4-Iodophenyl)-9H-carbazole

A448878-100MG A448878-1G A448878-250MG A448878-5G
76990-712EA 10 USD
76990-712 76990-714 76990-716 76990-718
9-(4-Iodophenyl)carbazole 98%
9-(4-Iodophenyl)carbazole

Precaución: for Research Use Only

Formula: C₁₈H₁₂IN
MDL Number: MFCD11617970
CAS Number: 57103-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR