1-Benzyl-4-(dimethylamino)piperidine 95%

Supplier: Ambeed
Warning

A448822-1G A448822-25G
77004-304EA 11 USD
77004-304 77004-306
1-Benzyl-4-(dimethylamino)piperidine 95%
1-Benzyl-4-(dimethylamino)piperidine

Precaución: for Research Use Only

Formula: C₁₄H₂₂N₂
MW: 218.33 g/mol
MDL Number: MFCD01084381
CAS Number: 64168-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR