1-[3,5-Bis(trifluoromethyl)phenyl]ethanamine Hydrochloride 97%

Supplier: Ambeed
Warning

A448770-1G A448770-250MG A448770-5G
77016-736EA 140.04 USD
77016-736 77016-738 77016-740
1-[3,5-Bis(trifluoromethyl)phenyl]ethanamine Hydrochloride 97%
1-[3,5-Bis(trifluoromethyl)phenyl]ethanamine Hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClF₆N
MW: 293.63 g/mol
MDL Number: MFCD04038875
CAS Number: 374822-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR