4,5-Dichloro-2,6-dimethylpyrimidine 95%

Supplier: Ambeed
Warning

A448488-1G A448488-250MG
77016-726EA 144.29 USD
77016-726 77016-728
4,5-Dichloro-2,6-dimethylpyrimidine 95%
4,5-Dichloro-2,6-dimethylpyrimidine

Precaución: for Research Use Only

Formula: C₆H₆Cl₂N₂
MW: 177.031 g/mol
MDL Number: MFCD16171717
CAS Number: 105742-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR