Methyl 3-methoxy-5-nitrobenzoate 97%

Supplier: Ambeed
Warning

A447431-1G A447431-250MG A447431-5G
76990-682EA 66 USD
76990-682 76990-684 76990-686
Methyl 3-methoxy-5-nitrobenzoate 97%
Methyl 3-methoxy-5-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₉NO₅
MDL Number: MFCD00157207
CAS Number: 78238-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR