Calcium 2-(trimethylammonio)ethyl phosphate chloride tetrahydrate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Phosphorylcholine chloride calcium salt tetrahydrate
A447277-10G
A447277-1G
A447277-250MG
A447277-5G
A447277-100G
A447277-1000G
A447277-25G
A447277-500G
76990-674EA
10.61
USD
76990-674
76990-676
76990-678
76990-680
77613-380
77684-227
77684-228
77684-229
Calcium 2-(trimethylammonio)ethyl phosphate chloride tetrahydrate 97%
Calcium 2-(trimethylammonio)ethyl phosphate chloride tetrahydrate
Precaución: For research use only.
Formula:
C₅H₂₁CaClNO₈P MW: 329.73 g/mol |
MDL Number:
MFCD00149983 CAS Number: 72556-74-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...