3-Bromo-2-methylbenzotrifluoride 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2-methyl-3-(trifluoromethyl)benzene

A446811-10G A446811-1G A446811-25G A446811-5G
77034-060EA 44.56 USD
77034-060 77034-062 77034-064 77034-066
3-Bromo-2-methylbenzotrifluoride 98%
3-Bromo-2-methylbenzotrifluoride

Precaución: for Research Use Only

Formula: C₈H₆BrF₃
MW: 239.04 g/mol
MDL Number: MFCD04038231
CAS Number: 69902-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR