5-Quinolinecarbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: Quinoline-5-carboxaldehyde, Quinoline-5-carbaldehyde

A446581-100MG A446581-1G A446581-250MG
77004-290EA 48.81 USD
77004-290 77004-292 77004-294
5-Quinolinecarbaldehyde 98%
5-Quinolinecarbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₇NO
MW: 157.17 g/mol
MDL Number: MFCD00805835
CAS Number: 22934-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR