1,2-Difluoro-4,5-diiodobenzene 98%

Supplier: Ambeed
Warning

A445763-100MG A445763-1G A445763-250MG
76990-668EA 63.66 USD
76990-668 76990-670 76990-672
1,2-Difluoro-4,5-diiodobenzene 98%
1,2-Difluoro-4,5-diiodobenzene

Precaución: for Research Use Only

Formula: C₆H₂F₂I₂
MW: 365.89 g/mol
MDL Number: MFCD24038778
CAS Number: 210567-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR