Zibotentan 97%

Supplier: Ambeed
Warning

Synonyms: N-(3-methoxy-5-methylpyrazin-2-yl)-2-[4-(1,3,4-oxadiazol-2-yl)phenyl]pyridine-3-sulphonamide, ZD4054, 2-(4-(1,3,4-Oxadiazol-2-yl)phenyl)-N-(3-methoxy-5-methylpyrazin-2-yl)pyridine-3-sulphonamide

A445755-10MG A445755-1MG A445755-25MG A445755-5MG
77016-718EA 196.27 USD
77016-718 77016-720 77016-722 77016-724
Zibotentan 97%
Zibotentan

Precaución: for Research Use Only

Formula: C₁₉H₁₆N₆O₄S
MW: 424.43 g/mol
MDL Number: MFCD09955343
CAS Number: 186497-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR