4-Bromo-N-isobutylaniline 95%

Supplier: Ambeed
Warning

A445727-100MG A445727-1G A445727-250MG A445727-5G
76990-662EA 18 USD
76990-662 76990-664 76990-666 77886-181
4-Bromo-N-isobutylaniline 95%
4-Bromo-N-isobutylaniline

Precaución: for Research Use Only

Formula: C₁₀H₁₄BrN
MDL Number: MFCD11144885
CAS Number: 195968-92-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR