1-Methyl-1H-indazole-4-carboxaldehyde 98%

Supplier: Ambeed
Warning

A445505-100MG A445505-1G A445505-250MG A445505-5G
77034-052EA 24 USD
77034-052 77872-725 77872-726 77872-727
1-Methyl-1H-indazole-4-carboxaldehyde 98%
1-Methyl-1H-indazole-4-carboxaldehyde

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MDL Number: MFCD10699167
CAS Number: 1053655-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR