N-(4-Methoxybenzyl)cyclopropanamine 97%

Supplier: Ambeed
Danger

A445498-1G A445498-5G A445498-25G
77016-714EA 48 USD
77016-714 77016-716 77890-901
N-(4-Methoxybenzyl)cyclopropanamine 97%
N-(4-Methoxybenzyl)cyclopropanamine

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO
MDL Number: MFCD07310991
CAS Number: 70894-71-2
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR