(1R,2R)-(-)-[1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene)]aluminum(III) chloride 98%

Supplier: Ambeed
Warning

Synonyms: (R,R)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminoaluminum chloride

A445113-100MG A445113-1G A445113-250MG A445113-5G
77016-700EA 21 USD
77016-700 77016-702 77016-704 77016-706
(1R,2R)-(-)-[1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene)]aluminum(III) chloride 98%
(1R,2R)-(-)-[1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene)]aluminum(III) chloride

Precaución: for Research Use Only

Formula: C₃₆H₅₂AlClN₂O₂
MDL Number: MFCD02684547
CAS Number: 250611-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR