(R)-(-)-1,1'-Binaphthyl-2,2'-diyl bis(trifluoromethanesulfonate) 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
(R)-(-)-1,1'-Binaphthol-2,2'-bis(trifluoromethanesulphonate), (R)-(-)-1,1'-Bi-2-naphthol Bis(trifluoromethanesulphonate), (R)-(-)-1,1'-Bi-2-naphthol bis(trifluoromethanesulphonate), (R)-[1,1'-Binaphthalene]-2,2'-diyl bis(trifluoromethanesulphonate)
A445007-1G
A445007-25G
A445007-5G
77004-280EA
7
USD
77004-280
77004-282
77004-284
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl bis(trifluoromethanesulfonate) 98%
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl bis(trifluoromethanesulfonate)
Precaución: for Research Use Only
Formula:
C₂₂H₁₂F₆O₆S₂ MW: 550.44 g/mol |
MDL Number:
MFCD00274615 CAS Number: 126613-06-7 UN: 3261 ADR: 8 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...