2,3-Dihydro-1H-pyrrolo[3,2-b]pyridine 97%

Supplier: Ambeed
Warning

A444946-100MG A444946-1G A444946-5G A444946-250MG
76990-652EA 126.25 USD
76990-652 76990-654 76990-656 77613-366
2,3-Dihydro-1H-pyrrolo[3,2-b]pyridine 97%
2,3-Dihydro-1H-pyrrolo[3,2-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₈N₂
MW: 120.15 g/mol
MDL Number: MFCD18256419
CAS Number: 1211540-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR