Biphenyl-3,4'-dicarboxylic acid 98%

Supplier: Ambeed
Warning

A444231-1G A444231-250MG
77016-694EA 49 USD
77016-694 77016-696
Biphenyl-3,4'-dicarboxylic acid 98%
Biphenyl-3,4'-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀O₄
MDL Number: MFCD00728324
CAS Number: 92152-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR