Biotin-PEG3-Amine 95%

Supplier: Ambeed
Warning

Synonyms: N-[2-[2-[2-(2-Aminoethoxy)ethoxy]ethoxy]ethyl]biotinamide, N-(2-(2-(2-(2-Aminoethoxy)ethoxy)ethoxy)ethyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide, N-Biotinyl-3,6,9-trioxaundecane-1,11-diamine, Amine-PEG3-Biotin

A444152-250MG A444152-1G A444152-100MG A444152-50MG
76990-646EA 216.43 USD
76990-646 77551-338 77601-920 77684-219
Biotin-PEG3-Amine 95%
Biotin-PEG3-amine

Precaución: For research use only.

Formula: C₁₈H₃₄N₄O₅S
MW: 418.55 g/mol
MDL Number: MFCD09952640
CAS Number: 359860-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR