Methyl 2-amino-5-chloro-3-nitrobenzoate 95%

Supplier: Ambeed
Warning

A444098-100MG A444098-1G A444098-250MG
76990-640EA 14 USD
76990-640 76990-642 76990-644
Methyl 2-amino-5-chloro-3-nitrobenzoate 95%
Methyl 2-amino-5-chloro-3-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₇ClN₂O₄
MDL Number: MFCD02068568
CAS Number: 84228-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR