8-Benzyl-2,8-diazaspiro[4.5]decan-3-one 98%

Supplier: Ambeed
Warning

A443839-1G A443839-250MG A443839-5G
76990-634EA 112.46 USD
76990-634 76990-636 76990-638
8-Benzyl-2,8-diazaspiro[4.5]decan-3-one 98%
8-Benzyl-2,8-diazaspiro[4.5]decan-3-one

Precaución: for Research Use Only

Formula: C₁₅H₂₀N₂O
MW: 244.332 g/mol
MDL Number: MFCD15147047
CAS Number: 154495-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR