Methyl benzo[d]thiazole-5-carboxylate 95+%

Supplier: Ambeed
Warning

A443731-100MG A443731-1G A443731-250MG A443731-5G
76990-626EA 54.11 USD
76990-626 76990-628 76990-630 76990-632
Methyl benzo[d]thiazole-5-carboxylate 95+%
Methyl benzo[d]thiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇NO₂S
MW: 193.22 g/mol
MDL Number: MFCD13176732
CAS Number: 478169-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR