5-Bromo-3,7-dimethyl-1H-indazole 95%
Supplier: Ambeed
|
Warning
|
A443566-100MG
A443566-1G
A443566-250MG
77034-046EA
46.68
USD
77034-046
77034-048
77034-050
5-Bromo-3,7-dimethyl-1H-indazole 95%
5-Bromo-3,7-dimethyl-1H-indazole
Precaución: for Research Use Only
Formula:
C₉H₉BrN₂ MW: 225.0900 g/mol |
MDL Number:
MFCD12755762 CAS Number: 1031417-71-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...