1-(4-Bromophenyl)cyclobutanecarboxylic acid 95%

Supplier: Ambeed
Warning

A442309-1G A442309-250MG A442309-5G
77034-028EA 22.28 USD
77034-028 77034-030 77572-380
1-(4-Bromophenyl)cyclobutanecarboxylic acid 95%
1-(4-Bromophenyl)cyclobutanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrO₂
MW: 255.1078 g/mol
MDL Number: MFCD08443165
CAS Number: 151157-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR