tert-Butyl 3-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate 98%

Supplier: Ambeed
Warning

A441802-1G A441802-250MG A441802-25G A441802-5G
76990-610EA 58.35 USD
76990-610 76990-612 76990-614 76990-616
tert-Butyl 3-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate 98%
tert-Butyl 3-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₃
MW: 227.30 g/mol
MDL Number: MFCD13568915
CAS Number: 478837-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR