3-Bromo-2-chloro-4-methyl-5-nitropyridine 98%
Supplier: Ambeed
|
Warning
|
A441685-100MG
A441685-1G
A441685-250MG
A441685-25G
A441685-5G
76990-604EA
21.22
USD
76990-604
76990-606
76990-608
77684-214
77684-215
3-Bromo-2-chloro-4-methyl-5-nitropyridine 98%
3-Bromo-2-chloro-4-methyl-5-nitropyridine
Precaución: For research use only.
Formula:
C₆H₄BrClN₂O₂ MW: 251.46 g/mol |
MDL Number:
MFCD12024452 CAS Number: 1049706-73-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...