Methyl 3-hydroxy-2-methylpropanoate 97%

Supplier: Ambeed
Warning

A441675-1G A441675-25G A441675-5G
76990-600EA 25 USD
76990-600 76990-602 77890-897
Methyl 3-hydroxy-2-methylpropanoate 97%
Methyl 3-hydroxy-2-methylpropanoate

Precaución: for Research Use Only

Formula: C₅H₁₀O₃
MDL Number: MFCD12194512
CAS Number: 42998-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR