5,6-Dimethylnicotinic acid 97%

Supplier: Ambeed
Warning

A441010-100MG A441010-250MG A441010-1G
77016-676EA 41 USD
77016-676 77016-678 77890-896
5,6-Dimethylnicotinic acid 97%
5,6-Dimethylnicotinic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MDL Number: MFCD13181797
CAS Number: 757903-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR