4-Chloro-6-methyl-1,3,5-triazin-2-amine 95%

Supplier: Ambeed
Warning

A440912-100MG A440912-1G
76990-594EA 59.42 USD
76990-594 76990-596
4-Chloro-6-methyl-1,3,5-triazin-2-amine 95%
4-Chloro-6-methyl-1,3,5-triazin-2-amine

Precaución: for Research Use Only

Formula: C₄H₅ClN₄
MDL Number: MFCD01566735
CAS Number: 21320-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR