(3aS,3a'S,8aR,8a'R)-2,2'-(Cyclopropane-1,1-diyl)bis(8,8a-dihydro-3aH-indeno[1,2-d]oxazole) ≥98%, ee 99%
Supplier: Ambeed
|
Warning
|
A440758-100MG
A440758-1G
A440758-250MG
A440758-5G
76990-586EA
42
USD
76990-586
76990-588
76990-590
76990-592
(3aS,3a'S,8aR,8a'R)-2,2'-(Cyclopropane-1,1-diyl)bis(8,8a-dihydro-3aH-indeno[1,2-d]oxazole) ≥98%, ee 99%
(3aS,3a'S,8aR,8a'R)-2,2'-(Cyclopropane-1,1-diyl)bis(8,8a-dihydro-3aH-indeno[1,2-d]oxazole)
Precaución: for Research Use Only
Formula:
C₂₃H₂₀N₂O₂ |
MDL Number:
MFCD29919664 CAS Number: 182122-08-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...