2-(3-Fluoro-5-nitrophenyl)acetic acid 98%

Supplier: Ambeed
Warning

A440304-100MG A440304-1G A440304-250MG
77034-016EA 33 USD
77034-016 77034-018 77034-020
2-(3-Fluoro-5-nitrophenyl)acetic acid 98%
2-(3-Fluoro-5-nitrophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₆FNO₄
MDL Number: MFCD13194392
CAS Number: 1211529-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR