4-Aminopyrimidine-5-carboxaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 4-Aminopyrimidine-5-carbaldehyde

A440076-100MG A440076-1G
77034-014EA 47.75 USD
77034-014 77568-876
4-Aminopyrimidine-5-carboxaldehyde 95%
4-Aminopyrimidine-5-carboxaldehyde

Precaución: For research use only.

Formula: C₅H₅N₃O
MW: 123.115 g/mol
MDL Number: MFCD09991893
CAS Number: 16357-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR