Boc-(R)-3-Cyclopentylalanine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-D-3-Cyclopentyl-Ala-OH, (R)-2-((tert-Butoxycarbonyl)amino)-3-cyclopentylpropanoic acid
A439736-100MG
A439736-1G
77016-672EA
44.56
USD
77016-672
77684-212
Boc-(R)-3-Cyclopentylalanine 95%
Boc-(R)-3-Cyclopentylalanine
Precaución: For research use only.
Formula:
C₁₃H₂₃NO₄ MW: 257.326 g/mol |
MDL Number:
MFCD03094882 CAS Number: 219819-74-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...