4-Amino-p-terphenyl 98%

Supplier: Ambeed
Warning

Synonyms: [1,1:4,1-Terphenyl]-4-amine

A439250-1G A439250-5G A439250-25G
77034-002EA 15 USD
77034-002 77034-004 77594-374
4-Amino-p-terphenyl 98%
4-Amino-p-terphenyl

Precaución: for Research Use Only

Formula: C₁₈H₁₅N
MW: 245.33 g/mol
MDL Number: MFCD00191302
CAS Number: 7293-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR