(4,5-Dichloro-2-methoxyphenyl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4,5-Dichloro-2-methoxyphenylboronic acid

A438310-1G A438310-250MG A438310-5G
77016-666EA 190.97 USD
77016-666 77016-668 77016-670
(4,5-Dichloro-2-methoxyphenyl)boronic acid 97%
(4,5-Dichloro-2-methoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BCl₂O₃
MW: 220.85 g/mol
MDL Number: MFCD17926484
CAS Number: 1256354-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR