5,5-Dioctyl-3,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5H-dibenzo[b,d]silole 95%

Supplier: Ambeed
Warning

A438225-100MG A438225-1G
77016-662EA 161.26 USD
77016-662 77016-664
5,5-Dioctyl-3,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5H-dibenzo[b,d]silole 95%
5,5-Dioctyl-3,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5H-dibenzo[b,d]silole

Precaución: for Research Use Only

Formula: C₄₀H₆₄B₂O₄Si
MW: 658.64 g/mol
MDL Number: MFCD16621135
CAS Number: 958293-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR